Skip to Content
Merck
All Photos(1)

Key Documents

182958

Poly(vinyl chloride-co-vinyl acetate)

powder

Synonym(s):

Pevikon C870

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH(Cl)-]x[-CH2CH(O2CCH3)-]y
CAS Number:
MDL number:
Pricing and availability is not currently available.

form

powder

autoignition temp.

725 °F

density

1.36 g/mL at 25 °C (lit.)

SMILES string

ClC=C.CC(=O)OC=C

InChI

1S/C4H6O2.C2H3Cl/c1-3-6-4(2)5;1-2-3/h3H,1H2,2H3;2H,1H2

InChI key

HGAZMNJKRQFZKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Warning

May contain vinyl chloride, an OSHA carcinogen. See 29 CFR part 1910

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service