Skip to Content
Merck
All Photos(1)

Key Documents

171530

Sigma-Aldrich

4-Phenylbutyronitrile

99%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
₹10,835.83

₹10,835.83


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
₹10,835.83

About This Item

Linear Formula:
C6H5(CH2)3CN
CAS Number:
Molecular Weight:
145.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

₹10,835.83


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.5143 (lit.)

bp

97-99 °C/1.7 mmHg (lit.)

density

0.973 g/mL at 25 °C (lit.)

functional group

nitrile
phenyl

SMILES string

N#CCCCc1ccccc1

InChI

1S/C10H11N/c11-9-5-4-8-10-6-2-1-3-7-10/h1-3,6-7H,4-5,8H2

InChI key

ICMVGKQFVMTRLB-UHFFFAOYSA-N

Application

4-Phenylbutyronitrile was used in the synthesis of light aliphatic nitriles and styrene derivatives[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic degradation of acrylonitrile-butadiene-styrene into fuel oil 2. Changes in the structure and catalytic activity of iron oxides.
Energy and Fuels, 15(3), 565-570 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service