Skip to Content
Merck
All Photos(1)

Key Documents

16603

Sigma-Aldrich

O-Methyl-undecaethylene glycol

≥95% (oligomer purity)

Synonym(s):

Methoxyundecaethylene glycol, Monomethoxy-PEG (n=11)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)11CH3
CAS Number:
Molecular Weight:
516.62
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (oligomer purity)

form

liquid

reaction suitability

reagent type: cross-linking reagent

SMILES string

COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C23H48O12/c1-25-4-5-27-8-9-29-12-13-31-16-17-33-20-21-35-23-22-34-19-18-32-15-14-30-11-10-28-7-6-26-3-2-24/h24H,2-23H2,1H3

InChI key

VWDQSWKLHABGKL-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Brero et al.
Nanomaterials (Basel, Switzerland), 10(9) (2020-08-28)
We present a 1H Nuclear Magnetic Resonance (NMR) relaxometry experimental investigation of two series of magnetic nanoparticles, constituted of a maghemite core with a mean diameter dTEM = 17 ± 2.5 nm and 8 ± 0.4 nm, respectively, and coated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service