Skip to Content
Merck
All Photos(1)

Key Documents

15578

Sigma-Aldrich

Borane 1,2-bis(tert-butylthio)ethane complex

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H28B2S2
CAS Number:
Molecular Weight:
234.08
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥97.0%

reaction suitability

reagent type: reductant

mp

~77 °C (dec.)

storage temp.

2-8°C

SMILES string

BB.CC(C)(C)SCCSC(C)(C)C

Other Notes

Nonflammable, stable, solid borane complex[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B.S. Kirkiacharian et al.
Synthesis, 383-383 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service