About This Item
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.6065 (lit.)
bp
128-129 °C/12 mmHg (lit.)
density
1.348 g/mL at 25 °C (lit.)
functional group
chloro
thioether
storage temp.
2-8°C
SMILES string
ClCSc1ccc(Cl)cc1
InChI
1S/C7H6Cl2S/c8-5-10-7-3-1-6(9)2-4-7/h1-4H,5H2
InChI key
XPJUCMIJGVAEGF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
No data available
Flash Point(C)
No data available
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service