Skip to Content
Merck
All Photos(1)

Key Documents

135283

Sigma-Aldrich

4-sec-Butylphenol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)C6H4OH
CAS Number:
Molecular Weight:
150.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

form

solid

bp

135-136 °C/25 mmHg (lit.)

mp

46-59 °C (lit.)

SMILES string

CCC(C)c1ccc(O)cc1

InChI

1S/C10H14O/c1-3-8(2)9-4-6-10(11)7-5-9/h4-8,11H,3H2,1-2H3

InChI key

ZUTYZAFDFLLILI-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Application

4-sec-Butylphenol was used in biotransformation of 4-sec-Butylphenol by hydrocarbon utilizing bacteria Mycobacterium neoaurum and Nocardia cyriacigeorgica.

Biochem/physiol Actions

4-sec-Butylphenol has estrogenic properties. It induces the CYP102 (Cytochrome P450BM-3) in Bacillus megaterium.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veronika Hahn et al.
Applied microbiology and biotechnology, 97(18), 8329-8339 (2013-08-06)
The environmental pollutant 4-sec-butylphenol (4-sec-BP) which possesses estrogenic properties was transformed by the aerobic Gram-positive bacteria Mycobacterium neoaurum and Nocardia cyriacigeorgica into three main products (P1-P3) which were isolated and structurally characterized in detail. Two of them were products of
N E Hopkins et al.
Biochemical and biophysical research communications, 244(3), 868-872 (1998-04-16)
CYP102 (Cytochrome P450BM-3) is induced in Bacillus megaterium by barbiturates, peroxisome proliferators, and nonsteroidal anti-inflammatory drugs. We now describe the induction of CYP102 in B. megaterium by 17 beta-estradiol and by 4-sec-butylphenol. These estrogens interact with the repressor protein Bm3R1
Gabino Bolívar-Subirats et al.
Environmental science and pollution research international, 27(33), 41314-41325 (2020-07-18)
The aim of this study was to develop a high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS)-based method for the multiresidue analysis of 21 plastic additives in river water. Analysed compounds included phthalates, benzophenone, bisphenol A and long- and short-chain alkylphenols (APs)
Markus A Lill et al.
Journal of medicinal chemistry, 48(18), 5666-5674 (2005-09-02)
We investigated the influence of induced fit of the androgen receptor binding pocket on free energies of ligand binding. On the basis of a novel alignment procedure using flexible docking, molecular dynamics simulations, and linear-interaction energy analysis, we simulated the
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service