Skip to Content
Merck
All Photos(1)

Documents

115886

Sigma-Aldrich

3-Chlorobenzyl chloride

98%

Synonym(s):

3,α-Dichlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2Cl
CAS Number:
Molecular Weight:
161.03
Beilstein:
742266
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.555 (lit.)

bp

215-216 °C (lit.)

density

1.27 g/mL at 25 °C (lit.)

SMILES string

ClCc1cccc(Cl)c1

InChI

1S/C7H6Cl2/c8-5-6-2-1-3-7(9)4-6/h1-4H,5H2

InChI key

DDGRAFHHXYIQQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Chlorobenzyl chloride has been used in the reaction of 3-methoxybenzyl chloride and ethyl 4-bromobenzoate in pure water, using zinc dust and a Pd catalyst.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Christophe Duplais et al.
Chemical communications (Cambridge, England), 46(4), 562-564 (2010-01-12)
A remarkably simple entry to unsymmetrical diarylmethanes has been developed that relies on an in situ organozinc-mediated, palladium-catalyzed cross-coupling. Thus, by mixing a benzyl and aryl halide together in the presence of Zn metal and a Pd catalyst, diarylmethanes are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service