All Photos(1)
About This Item
Linear Formula:
ONC10H6OH
CAS Number:
Molecular Weight:
173.17
Colour Index Number:
10010
Beilstein:
1867919
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
95%
mp
150-155 °C (dec.) (lit.)
SMILES string
Oc1c(ccc2ccccc12)N=O
InChI
1S/C10H7NO2/c12-10-8-4-2-1-3-7(8)5-6-9(10)11-13/h1-6,12H
InChI key
SYUYTOYKQOAVDW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
2-nitroso-1-naphthol helps in the extraction of cobalt ion as it is used as a selective complexing agent. It is used in the separation and preconcentration of Th(IV), Ti(IV), Fe(III), Pb(II) and Cr(III) by solid phase extraction.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Shiho Ohnishi et al.
Japanese journal of cancer research : Gann, 93(7), 736-743 (2002-08-01)
2-Naphthylamine (2-NA), a bladder carcinogen, is contained in cigarette smoke. DNA adduct formation is thought to be a major cause of DNA damage by carcinogenic aromatic amines. We have investigated whether a metabolite of 2-NA, 2-nitroso-1-naphthol (NO-naphthol) causes oxidative DNA
Mahmoud Chamsaz et al.
Environmental monitoring and assessment, 185(11), 9067-9075 (2013-05-08)
Highly rapid and selective vortex-assisted liquid-liquid microextraction based on solidification of organic drop has been used for determination of cobalt ion. 2-Nitroso-1-naphthol (2N1N) was used as a selective complexing agent to form stable cobalt-2N1N complex which can be extracted with
Funda Armagan Aydin et al.
Journal of hazardous materials, 173(1-3), 669-674 (2009-09-22)
A simple and effective method is presented for the separation and preconcentration of Th(IV), Ti(IV), Fe(III), Pb(II) and Cr(III) by solid phase extraction on 2-nitroso-1-naphthol impregnated MCI GEL CHP20P resin prior to their inductively coupled plasma-mass spectrometric determinations. The influence
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service