Skip to Content
Merck
All Photos(1)

Key Documents

101648

Sigma-Aldrich

3-Chloro-4-methylaniline

99%

Synonym(s):

3-Chloro-p-toluidine, 4-Amino-2-chlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)NH2
CAS Number:
Molecular Weight:
141.60
Beilstein:
636511
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

99%

refractive index

n20/D 1.584 (lit.)

bp

237-238 °C (lit.)

mp

24-25 °C (lit.)

density

1.167 g/mL at 25 °C

SMILES string

Cc1ccc(N)cc1Cl

InChI

1S/C7H8ClN/c1-5-2-3-6(9)4-7(5)8/h2-4H,9H2,1H3

InChI key

RQKFYFNZSHWXAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

DuPont product

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yesol Bak et al.
Journal of microbiology and biotechnology, 28(4), 645-651 (2018-03-16)
The carcinogenicity of chemicals in the environment is a major concern. Recently, numerous studies have attempted to develop methods for predicting carcinogenicity, including rodent and cell-based approaches. However, rodent carcinogenicity tests for evaluating the carcinogenic potential of a chemical to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service