Skip to Content
Merck
All Photos(1)

Key Documents

AABH99A46CF6

AA BLOCKS, INC.

ethyl 6-hydroxy-1,3-benzothiazole-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
223.25
MDL number:

Iupac Name

ethyl 6-hydroxy-1,3-benzothiazole-2-carboxylate

Inchi Code

1S/C10H9NO3S/c1-2-14-10(13)9-11-7-4-3-6(12)5-8(7)15-9/h3-5,12H,2H2,1H3

InChI key

NKWMUUYSJMDDID-UHFFFAOYSA-N

storage temp.

2~8 C

Country of Origin

US

Product Link

https://www.aablocks.com/prod/59587-30-3

Synonym

ethyl 6-hydroxy-1,3-benzothiazole-2-carboxylate

MSDS

https://www.aablocks.com/downloadSds?cas=59587-30-3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library