1,3-Butadiene solution
Empirical Formula (Hill Notation):
C4H6
CAS Number:
Molecular Weight:
54.09
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing