Skip to Content
Merck
All Photos(2)

Key Documents

V900611

Sigma-Aldrich

Dimethyl isophthalate

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
Beilstein:
1912251
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor density

6.8 (vs air)

vapor pressure

93.7 mmHg ( 208 °C)

product line

Vetec

Assay

98%

mp

64-68 °C (lit.)

SMILES string

COC(=O)c1cccc(c1)C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-4-3-5-8(6-7)10(12)14-2/h3-6H,1-2H3

InChI key

VNGOYPQMJFJDLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

343.4 °F - closed cup

Flash Point(C)

173 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service