Skip to Content
Merck
All Photos(1)

Documents

V900358

Sigma-Aldrich

6-(γ,γ-Dimethylallylamino)purine

Vetec, reagent grade, ≥98.5%

Synonym(s):

2iP, N6-(2-Isopentenyl)adenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5
CAS Number:
Molecular Weight:
203.24
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥98.5%

application(s)

agriculture

storage temp.

−20°C

SMILES string

C\C(C)=C\CNc1ncnc2[nH]cnc12

InChI

1S/C10H13N5/c1-7(2)3-4-11-9-8-10(13-5-12-8)15-6-14-9/h3,5-6H,4H2,1-2H3,(H2,11,12,13,14,15)

InChI key

HYVABZIGRDEKCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Ishii et al.
Biochimica et biophysica acta, 1745(2), 156-165 (2005-09-01)
Cytokinins are important purine derivatives that act as redifferentiation-inducing hormones to control many processes in plants. Cytokinins such as isopentenyladenine (IPA) and kinetin are very effective at inducing the granulocytic differentiation of human myeloid leukemia HL-60 cells. We examined the
Ming-Tang Chen et al.
Nature biotechnology, 23(12), 1551-1555 (2005-11-22)
The construction of synthetic cell-cell communication networks can improve our quantitative understanding of naturally occurring signaling pathways and enhance our capabilities to engineer coordinated cellular behavior in cell populations. Towards accomplishing these goals in eukaryotes, we developed and analyzed two
A Azmi et al.
Planta, 213(1), 29-36 (2001-08-29)
Our previous results demonstrated that endogenous cytokinins are involved in the shooty potential of tumors initiated on Eucalyptus globulus plantlets inoculated with Agrobacterium tumefaciens strain 82.139 [A. Azmi et al. (1997a) Plant Sci 127: 81-90]. In order to investigate whether
Yuki Ishii et al.
British journal of haematology, 128(4), 540-547 (2005-02-03)
Cytokinins, purine derivatives that act as hormones to control many processes in plants, are very effective at inducing the granulocytic differentiation of human myeloid leukaemia cells. Isopentenyladenine (IPA), a potent cytokinin, significantly induced the expression of CCAAT/enhancer-binding protein (C/EBP)delta, but
Patrick von Aderkas et al.
Tree physiology, 22(6), 431-434 (2002-04-19)
Embryogenic tissue of hybrid larch (Larix x marschlinsii Coaz) was multiplied on Medium M (modified MSG medium supplemented with the plant growth regulators (PGRs) 2,4-dichlorophenoxyacetic acid (2,4-D; 9 microM) and N-6-benzyladenine (2.25 microM)). After 1 week, cultures were transferred to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service