Skip to Content
Merck
All Photos(1)

Documents

1581504

USP

Pseudoephedrine sulfate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(1S,2S)-(+)-Pseudoephedrine sulfate salt, α-(1-Methylaminoethyl)benzyl alcohol sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32N2O6S
CAS Number:
Molecular Weight:
428.54
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

pseudoephedrine

manufacturer/tradename

USP

mp

175-179 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OS(O)(=O)=O.CN[C@@H](C)[C@@H](O)c1ccccc1.CN[C@@H](C)[C@@H](O)c2ccccc2

InChI

1S/2C10H15NO.H2O4S/c2*1-8(11-2)10(12)9-6-4-3-5-7-9;1-5(2,3)4/h2*3-8,10-12H,1-2H3;(H2,1,2,3,4)/t2*8-,10+;/m00./s1

InChI key

CAVQBDOACNULDN-NRCOEFLKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pseudoephedrine sulfate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Brompheniramine Maleate and Pseudoephedrine Sulfate Oral Solution
  • Capsules Containing at Least Three of the Following—Acetaminophen and Salts of Chlorpheniramine, Dextromethorphan, and Pseudoephedrine
  • Dexbrompheniramine Maleate and Pseudoephedrine Sulfate Oral Solution
  • Ephedrine Sulfate Injection
  • Oral Powder Containing at Least Three of the Following—Acetaminophen and Salts of Chlorpheniramine, Dextromethorphan, and Pseudoephedrine
  • Oral Solution Containing at Least Three of the Following—Acetaminophen and Salts of Chlorpheniramine, Dextromethorphan, and Pseudoephedrine
  • Pseudoephedrine Sulfate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service