Skip to Content
Merck
All Photos(1)

Key Documents

PHL22330

Melatonin

new

phyproof® Reference Substance

Synonym(s):

Circadin, Melatol, Melatonine, Melaxen, Melovine, Regulin, Slenyto, N-Acetyl-5-methoxytryptamine

Sign Into View Organizational & Contract Pricing

Select a Size

PHL22330-50MG
₪1,507.00

₪1,507.00


Please contact Customer Service for Availability


Select a Size

Change View
PHL22330-50MG
₪1,507.00

About This Item

Empirical Formula (Hill Notation):
C13H16N2O2
CAS Number:
Molecular Weight:
232.28
Beilstein:
205542
MDL number:
UNSPSC Code:
12352200

₪1,507.00


Please contact Customer Service for Availability

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

technique(s)

HPLC: suitable

mp

116.5-118 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc2[nH]cc(CCNC(C)=O)c2c1

InChI

1S/C13H16N2O2/c1-9(16)14-6-5-10-8-15-13-4-3-11(17-2)7-12(10)13/h3-4,7-8,15H,5-6H2,1-2H3,(H,14,16)

InChI key

DRLFMBDRBRZALE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service