G005676
HPLC Analysis of Benzoic Acid Derivatives on Ascentis® C18
application for HPLC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
technique(s)
HPLC: suitable
test parameters
column: Ascentis C18, 15 cm x 4.6 mm I.D., 5 μm particles (581324-U)
mobile phase: [A] 10mM TFA in water; [B] 10 mM TFA in acetonitrile; (70:30, A:B)
flow rate: 1.0 mL/min
column temp.: 30 °C
detector: UV, 220 nm
injection: 10 μL
suitability
application for HPLC
application(s)
vitamins, nutraceuticals, and natural products
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Analyte
Description
4-Hydroxybenzoic acid
Acetylsalicylic acid
Benzoic acid, meets USP testing specifications
Salicylic acid, puriss. p.a., ≥99.0% (T)
Ethyl 4-hydroxybenzoate, ReagentPlus®, 99%
analytical column
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service