Recommended Products
grade
reference material
Quality Level
packaging
pkg of 50 mL
manufacturer/tradename
Fera Science Ltd
application(s)
food and beverages
format
matrix material
storage temp.
2-8°C
Related Categories
Other Notes
Certified for the analytes listed below. See certificate for values and more details
Trace Elements
Cadmium (Cd), Copper (Cu), Lead (Pb)
Matrix Group: Beverages and Water
Trace Elements
Cadmium (Cd), Copper (Cu), Lead (Pb)
Matrix Group: Beverages and Water
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service