Skip to Content
Merck
All Photos(1)

Key Documents

808807

TSKgel® SP-5PW (20 µm) HPLC Columns

L x I.D. 1 cm × 8 mm, sulfonic acid phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
₪4,787.00

₪4,787.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
₪4,787.00

About This Item

UNSPSC Code:
41115711

₪4,787.00


Please contact Customer Service for Availability

Product Name

TSKgel® SP-5PW Guardgel Kit, phase sulfonic acid, L × I.D. 1 cm × 8 mm, 20 μm particle size, glass

material

glass

product line

TSKgel®

feature

counter ion Na+

manufacturer/tradename

Tosoh 08807

parameter

≤1 M salt concentration
≤20% organic solvent
10-45 °C temperature

technique(s)

LPLC: suitable

L × I.D.

1 cm × 8 mm

matrix

hydroxylated methacrylic polymer

matrix active group

sulfonic acid phase

particle size

20 μm

operating pH

2-12

separation technique

cation exchange

General description

Each TSKgel® Guardgel Kit contains 5 mL packing material, specified in the product description, in an appropriate slurry solvent. Guardgel Kits to protect glass columns also include a column holder, a glass column, 10 mm L x 13 mm OD x 8 mm ID, 2 external endfittings, 2 caps with M6 threads, and a packing adaptor. Simple instructions to pack the guard column are included in the shipment.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service