Skip to Content
Merck
All Photos(1)

Documents

58701

PTFE Tubing

L × O.D. × I.D. 10 ft × 1/16 in. × 0.023 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

packaging

pkg of 1 ea

L × O.D. × I.D.

10 ft × 1/16 in. × 0.023 in.

Looking for similar products? Visit Product Comparison Guide

General description

Economical, flexible PTFE tubing is ideal for use at pressures up to 500 psi (35 kg/cm2). Use in automatic analyzer, postcolumn reaction, preparative scale systems, in-stream sampling devices, and when monitoring physiologically important compounds.

The maximum recommended operating temperature for PTFE is 200 °C, but short term exposure to higher temperatures seldom causes damage.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keunhong Jeong et al.
Magnetic resonance in chemistry : MRC, 56(11), 1089-1093 (2018-06-02)
Parahydrogen is a potentially significant source of hyperpolarization. However, a heat exchanger at an ultralow temperature, which is normally sustained wastefully using liquid nitrogen, is essential for the generation of hyperpolarized parahydrogen. In order to cut down on the use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service