Skip to Content
Merck
All Photos(1)

Documents

48078

Supelco

1,3-Dichloro-2-propanol solution

certified reference material, 2000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6Cl2O
CAS Number:
Molecular Weight:
128.99
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C3H6Cl2O/c4-1-3(6)2-5/h3,6H,1-2H2

InChI key

DEWLEGDTCGBNGU-UHFFFAOYSA-N

Application

1,3-Dichloro-2-propanol may be used as a certified reference standard for the quantification of the analyte in papers treated with polyamidoamine-epichlorohydrin wet-strength resins using gas chromatography coupled to mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 1, 3-dichloro-2-propanol and 3-chloro-1, 2-propanediol in papers treated with polyamidoamine-epichlorohydrin wet-strength resins by gas chromatography-mass spectrometry using selective ion monitoring
Boden L, et al.
Journal of Chromatography A, 788(1-2), 195-203 (1997)
Diomi Mamma et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 41(3), 303-313 (2006-02-18)
The removal of 1,3-dichloro-2-propanol (1,3-DCP), 3-chloro-1,2-propanediol (3-CPD) and their mixtures at concentrations up to 1,000 mg . L(-1) by the whole cell system of Pseudomonas putida DSM 437 was investigated. The 1,3-DCP removal rates ranged from 2.36 to 10.55 mg
Sarah Hasnip et al.
Journal of AOAC International, 88(5), 1404-1412 (2006-01-03)
An interlaboratory study was performed to evaluate the effectiveness of a headspace gas chromatography (GC) method for the determination of 1,3-dichloro-propan-2-ol (1,3-DCP) in soy sauce and related products at levels above 5 ng/g. The test portion is mixed with an
Rainer Schuhmacher et al.
Analytical and bioanalytical chemistry, 382(2), 366-371 (2005-04-28)
A rapid analytical method for sensitive determination of 1,3-dichloro-2-propanol (1,3-DCP) in river water has been developed. 1,3-DCP is extracted from water with ethyl acetate. After filtration through sodium sulfate the ethyl acetate phase is analyzed by gas chromatography-mass spectrometry. The
Hyeon-Yeong Kim et al.
The Annals of occupational hygiene, 51(7), 633-643 (2007-10-09)
The subchronic toxicity of 1,3-dichloro-2-propanol (1,3-DCP) was investigated in Fischer 344 rats after 13 weeks of repeated, whole-body inhalation exposure. Groups of 10 rats of each sex were exposed to 1,3-DCP vapor by whole-body inhalation exposure at concentrations of 0

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service