Skip to Content
Merck
All Photos(29)

Documents

ZRB1172

Sigma-Aldrich

Anti-S6 Ribosomal Protein Antibody, clone 1E9, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E9, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 28.68 kDa
observed mol wt ~32 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable (peptide)

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RPS6(6194)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1E9 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects 40S ribosomal protein S6. It targets an epitope within the N-terminal region.

Immunogen

His-tagged recombinant fragment corresponding to 136 amino acids from the N-terminal region of human 40S Ribosomal Protein S6.

Application

Anti-S6 Ribosomal Protein, clone 1E9 ZooMAb , Cat. No. ZRB1172, is a highly specific rabbit recombinant monoclonal antibody that targets S6 Ribosomal Protein and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected S6 Ribosomal Protein in human pancreas and mouse pancreas tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected S6 Ribosomal Protein in HeLa, A431, HUVEC, NIH 3T3, Jurkat, MCF-7 cells.

Note: Actual optimal working dilutions must be determined by the end user as specimens and experimental conditions may vary.

Target description

40S ribosomal protein S6 (UniProt: P62753; also known Phosphoprotein NP33, Small ribosomal subunit protein eS6; RpS6) is encoded by the RPS6 (also known as OK/SW-cl.2) gene (Gene ID: 6194) in human. RpS6 is a highly conserved member of the eukaryotic ribosomal protein eS6 family and is a component of the 40S ribosomal subunit. It plays an important role in controlling cell growth and proliferation through the selective translation of particular classes of mRNA. It is the major substrate of protein kinases in eukaryote ribosomes and its phosphorylation is stimulated by growth factors, tumor promoting agents, and mitogens. RpS6 can be phosphorylated on five serine residues (Ser235, 236, 240, 244, and 247) in the region close to the carboxy terminus. Among the serine residues 235 and 236 are shown to be directly phosphorylated by S6Kinase that is subjected to regulation by the nutrient-mTORC1 signaling. Phosphorylation at serine 240/244 is reported to be predominantly nuclear but is also detectable in the cytoplasm, whereas serine 235/236 phosphorylated S6 is almost exclusively localized to the nucleus of primary human cells and is virtually undetectable in the cytoplasm, except in transformed cells where it can also be detected in the cytoplasm. RpS6 is also shown to exist in complex with the Kaposi s sarcoma-associated herpesvirus latency-associated nuclear antigen (LANA). This interaction is mediated by the N-terminal domain of LANA. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Rosner, M., et al. (2011). Amino Acids 40(2); 595-600; Chen, W., and Dittmar, DP (2011). J. Virol. 85(18); 9495-9505; Biever, A., et al. (2015). J. Neurosci. 2015, 35(10); 4113-4130)

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose. Normal appearance is a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bideep Shrestha et al.
Scientific reports, 14(1), 1066-1066 (2024-01-12)
Folate receptor (FR) alpha (FOLR1) and beta (FOLR2) are membrane-anchored folate transporters that are expressed at low levels in normal tissues, while their expression is strongly increased in several cancers. Intriguingly, although the function of these receptors in, for example

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service