Z688304
LightSafe micro centrifuge tubes
capacity 0.5 mL, conical black polypropylene
About This Item
Recommended Products
material
cap (attached)
conical black polypropylene
sterility
non-sterile
feature
RCF 15,000 × g
RNase and DNase free
packaging
pack of 500
manufacturer/tradename
Cole Parmer 633375
capacity
0.5 mL
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- RNase and DNase free tubes
- Opaque tubes protect light-sensitive compounds
- Used along with fluorescent probes or ethidium bromide.
- Made of biocompatible polypropylene
- The click-fit cap of the tube prevents accidental leakage while opening.
- Suitable for freezing to -100°C.
- Autoclavable at 122°C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service