Skip to Content
Merck
All Photos(1)

Key Documents

Z648078

Microcon centrifugal filter unit

YM-10 membrane, NMWCO 10 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103900

material

polycarbonate vial
polypropylene container
regenerated cellulose membrane
rubber O-ring

manufacturer/tradename

Millipore 4205
Millipore 4206
Millipore 42407

parameter

~0.5 mL sample volume

concentrate volume

5-15 μL

membrane area × diam. × L

0.32 cm2 × 12.3 mm × 45 mm

color

green

pore size

10 kDa NMWCO

Looking for similar products? Visit Product Comparison Guide

General description

The lab standard for sub-microliter volumes. Ideal for protein, nucleic acid concentration, desalting and buffer exchange or removal of primers, linkers and unincorporated labels.
Fixed-angle microcentrifuge rotor with max. 14,000 x g.
  • Solute recoveries typically >95%, ideal for dilute (ng.mL ranger) protein solutions
  • Has a deadstop for reproducible concentrate volumes
  • Inverted spin method of concentrate retrieval maximizes recovery
  • Low-binding Ultracel-YM membrane
  • Color coding indicates MW cut-off
  • Convenient sample storage of filtrate or concentrated sample in standard microcentrifuge collection tube
  • Use in standard 1.5 mL tube fixed-angle rotors

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service