Key Documents
Z615080
Vivapore 5 solvent absorption concentrator
MWCO 7.5 kDa
About This Item
UNSPSC Code:
41104916
NACRES:
NB.24
material
polyethersulfone membrane
styrene-acrylonitrile (SAN) housing
description
min concentrate vol 50 μL
sterility
non-sterile
manufacturer/tradename
Sartorius VP0502
availability
not available in USA and Canada, ROW
membrane area
20 cm2
working volume
5 mL
pore size
7.5 kDa MWCO
General description
With no need for additional equipment, pressure or vacuum, solvent absorption is the most economic and user friendly concentration technique available to the clinician and research scientist. Just fill the unit with the solution to be concentrated, wait for the desired concentration level to be achieved and then pipette the concentration sample from the bottom of the reservoir. Ideal for general purpose laboratory concentration or purification prior to further analysis, Vivapore is particularly suited for labile solutions that can denature with alternative shear or pressure inducing methods or require processing in a cold room environment. Vivapore concentrators extend the solvent absorption technique to a totally new level of performance, application potential and ease of use. Requires stand.
Choose from one of the most recent versions:
Certificates of Analysis (COA)