Skip to Content
Merck
All Photos(1)

Key Documents

Z316121

Eppendorf® PCR tubes

capacity 0.2 mL, colorless

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103037
NACRES:
NB.22

material

cap (Lid)
polypropylene

sterility

non-sterile

feature

RNase and DNase free

packaging

pkg of 1,000 tubes (bulk packed)

manufacturer/tradename

Eppendorf® 0030 124.332

availability

not available in USA and Canada

technique(s)

PCR: suitable

capacity

0.2 mL

color

colorless

Looking for similar products? Visit Product Comparison Guide

General description

Thin-wall PCR tubes with a contamination shield at the front edge of the lid to prevent accidental contact with the inner surface when the tube is opened.
Matt writing surface on lid

Chem/Phys Resistance

Autoclavable

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service