Skip to Content
Merck
All Photos(1)

Key Documents

T8819

Sigma-Aldrich

TFIIB human

~5 units/μL, recombinant, expressed in E. coli, solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 UNITS
₪2,407.00

₪2,407.00


Please contact Customer Service for Availability


Select a Size

Change View
50 UNITS
₪2,407.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

₪2,407.00


Please contact Customer Service for Availability

recombinant

expressed in E. coli

Quality Level

form

solution

mol wt

~32 kDa

concentration

~5 units/μL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... GTF2B(2959)

Biochem/physiol Actions

General transcription factor for RNA Polymerase II.
The amino terminal region of TFIIB contains a zinc ribbon fold composed of antiparallel Β strands. Mutational studies show that this domain is required for recruitment of RNA Pol II for transcriptional initiation. [1]

Unit Definition

One gel shift unit is the amount of TFIIB required to form 0.2 pmol of TFIIB/TFIID/DNA complex as detected by gel shift.

Physical form

Solution in 20 mM Tris-HCl, pH 7.4, 100 mM NaCl, 2 mM DTT, 1 mM EDTA, 0.1% Triton® X-100.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Hahn et al.
Genes & development, 14(6), 719-730 (2000-03-25)
The function of the conserved zinc-binding domains in the related Pol II- and Pol III-specific factors TFIIB and Brf was investigated. Three-dimensional structure modeling and mutagenesis studies indicated that for both factors, the functional surface of the zinc ribbon fold

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service