T1139
TNPAL-glucocerebroside
Synonym(s):
Trinitrophenylaminolauroylglucocerebroside
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C42H71N5O14
CAS Number:
Molecular Weight:
870.04
MDL number:
UNSPSC Code:
12352211
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCC\C=C\C(O)C(COC1OC(CO)C(O)C(O)C1O)NC(=O)CCCCCCCCCCCNc2c(cc(cc2N(=O)=O)N(=O)=O)N(=O)=O
Preparation Note
Prepared from glucocerebrosides from Human (Gaucher′s) Spleen
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service