Skip to Content
Merck
All Photos(1)

Documents

T1074

Sigma-Aldrich

Tricine

Biotechnology Performance Certified, 1 M in solution, suitable for cell culture, 0.2 μm filtered

Synonym(s):

N-[Tris(hydroxymethyl)methyl]glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)3CNHCH2CO2H
CAS Number:
Molecular Weight:
179.17
Beilstein:
1937804
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

grade

Biotechnology Performance Certified

sterility

0.2 μm filtered

concentration

1 M in solution

technique(s)

cell culture | mammalian: suitable

impurities

DNase, RNase, Protease, none detected
bioburden, tested
endotoxin, tested
<5 ppm heavy metals as (Pb)

useful pH range

7.4-8.8

pKa (25 °C)

8.1

cation traces

Fe: <5 ppm

SMILES string

OCC(CO)(CO)NCC(O)=O

InChI

1S/C6H13NO5/c8-2-6(3-9,4-10)7-1-5(11)12/h7-10H,1-4H2,(H,11,12)

InChI key

SEQKRHFRPICQDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Buffer component for separation of low molecular weight peptides.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arpita Gantayet et al.
Biofouling, 29(1), 77-85 (2012-12-06)
The freshwater zebra mussel (Dreissena polymorpha) is a notorious biofouling organism. It adheres to a variety of substrata underwater by means of a proteinaceous structure called the byssus, which consists of a number of threads with adhesive plaques at the
Syed R Haider et al.
Methods in molecular biology (Clifton, N.J.), 869, 81-91 (2012-05-16)
Tricine-sodium dodecyl sulphate-polyacrylamide gel electrophoresis (tricine-SDS-PAGE) is an efficient way of separating low-molecular-mass proteins. However, the standard system is quite complicated and specifically may not be useful when the separated proteins require to be recovered from the gel for quantitative
Lindsay M McCracken et al.
Neuropharmacology, 58(3), 676-681 (2009-11-17)
Glycine receptor function mediates most inhibitory neurotransmission in the brainstem and spinal cord and is enhanced by alcohols, volatile anesthetics, inhaled drugs of abuse, and endogenous compounds including zinc. Because zinc exists ubiquitously throughout the brain, investigations of its effects
Syed R Haider et al.
Journal of separation science, 34(18), 2463-2467 (2011-08-06)
This paper compares different buffer systems for the electrophoretic separation of the five most abundant serum proteins on native-PAGE gel and cellulose membranes. A modified Tris-tricine system was shown to be superior for the separation of these serum proteins in
Aleksandr Shcheglovitov et al.
The Journal of general physiology, 139(3), 219-234 (2012-03-01)
Here, we describe a new mechanism by which glutamate (Glu) and trace metals reciprocally modulate activity of the Ca(v)2.3 channel by profoundly shifting its voltage-dependent gating. We show that zinc and copper, at physiologically relevant concentrations, occupy an extracellular binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service