Skip to Content
Merck
All Photos(1)

Key Documents

SRP6297

Sigma-Aldrich

Cathepsin S human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

CTSS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

~37 kDa

packaging

pkg of 10 μg
pkg of 50 μg

concentration

0.2 μM

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CTSS(1520)

General description

Cathepsin S (CTSS) is a lysosomal cysteine endopeptidase of the papain family or C1 family. It is produced as an inactive preproenzyme which is then converted into the active form in lysosomes and late endosomes.[1][2]

Biochem/physiol Actions

Cathepsin S (CTSS) is a key participant in MHC (major histocompatibility complex) class II-mediated immune responses, where CTSS degrades Ii (type II membrane glycoprotein invariant chain) sequence of MHC II aβ heterodimers. Ii contains signaling motifs and its degradation leads to successful association of class II associated invariant chain peptide (CLIP) with the peptide groove of the MHC class II molecule. This is essential for antigen loading on MHC II complex during antigen presentation.[1] Overexpression of this protein in mice leads to increased proteinase-activated receptor-2 (PAR-2) expression in dendritic cells resulting in scratching behavior and Th1 (T-helper) cell-associated cytokine expression. This can eventually lead to chronic atopic dermatitis in mice.[2]

Physical form

A 0.2 μM filtered solution of 20mM MES, 150mM NaCl, 10% Glycerol, pH 5.5.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah F Hamm-Alvarez et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(7), 1872-1881 (2014-03-20)
The diagnosis of Sjögren's syndrome (SS) in routine practice is largely a clinical one and requires a high index of suspicion by the treating physician. This great dependence on clinical judgment frequently leads to delayed diagnosis or misdiagnosis. Tear protein
Nari Kim et al.
The Journal of investigative dermatology, 132(4), 1169-1176 (2011-12-16)
Atopic dermatitis (AD) is a chronically relapsing, non contagious pruritic skin disease with two phases: acute and chronic. Cysteine protease cathepsin S (CTSS) is involved in inflammatory processes, possibly leading to atherosclerosis and asthma. Recently, it has been reported that
Si-Tu Xue et al.
RSC advances, 9(15), 8600-8607 (2019-03-14)
Cathepsin K (Cat K) is a predominant cysteine protease and highly potent collagenase expressed in osteoclasts. Cat K inhibitors are anti-resorptive agents to treat osteoporosis. A novel scaffold of cathepsin K inhibitors, exemplified by lead compound 1x, was used as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service