Skip to Content
Merck
All Photos(2)

Key Documents

SRP6255

Sigma-Aldrich

VEGF-B human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

VEGFB, VEGFL, VRF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 20.2 kDa
observed mol wt 15-18 kDa (DTT-reduced. Protein migrates due to glycosylation. Arg 22 is the predicted N-terminus.)
observed mol wt 33-35 kDa

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VEGF-B(7423)

General description

VEGFB (vascular endothelial growth factor B) is encoded by the gene mapped to human chromosome 11q13. The encoded protein is one of the seven members of the VEGF family, which also includes VEGF-A, VEGF-C, VEGF-D, VEGF-E, VEGF-F, and PIGF (placental growth factor). All these members contain a common VEGF homology domain. This domain forms the core region and contains a cystine knot motif composed of eight invariant cysteine residues. VEGFB, discovered in 1995, is highly expressed in adult myocardium, skeletal muscle, and pancreas. This gene contains seven exons and alternative splicing results in the synthesis of two isoforms, VEGF-B167 and VEGF-B186, with molecular weight of 21kDa and 32kDa, respectively. These isoforms show differential level of expression, with VEGF-B167 being the predominantly expressed isoform.

Biochem/physiol Actions

VEGFB (vascular endothelial growth factor B) acts on the receptors VEGFR-1 (VEGF receptor) and NRP-1 (neuropilin). This protein induces coronary vessel growth and cardiac hypertrophy, which might confer protection to heart against ischemic damage and heart failure. It might have a role in metabolic functions as it shows high expression levels in tissues with highly active energy metabolism. VEGFB is also thought to be involved in neuroprotection. This protein is involved in fetal angiogenesis, functions as a mitogen for endothelial cells (ECs), survival factor for multiple types of cells such as neurons, vascular cells and myocytes.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roles of vascular endothelial growth factor in amyotrophic lateral sclerosis.
Pronto-Laborinho AC et al
BioMed Research International, 2014, 947513-947513 (2014)
Vascular endothelial growth factor and angiogenesis.
Hoeben A et al
Pharmacological Reviews, 56(4), 549-580 (2004)
Vascular endothelial growth factor-B in physiology and disease.
Bry M et al
Physiological Reviews, 94(3), 779-794 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service