Skip to Content
Merck
All Photos(1)

Documents

SRP6206

Sigma-Aldrich

sCD22 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

B Lymphocyte cell adhesion molecule, BL-CAM, Leu-14, Siglec-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

mol wt

~75 kDa

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CD22(933)

General description

CD22 is a B-lineage restricted 135 kDa glycoprotein whose cell surface expression is limited to resting and activated B lymphocytes. The physiological role of CD22 is still unknown. Targeted disruption of CD22 in mice results in a reduced level of surface IgM on peripheral B cells suggesting a role for CD22 in limiting antigen receptor signaling. CD22 is a member of the Ig gene superfamily that uniquely binds a sialic acid-dependent ligand. Recombinant human CD22 is a soluble 75.0 kDa protein which corresponds to the extracellular domain of CD22.

Physical form

Sterile filtered through a 0.2 micron filter. Lyophilized from 1X PBS, pH 7.2.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service