Skip to Content
Merck
All Photos(1)

Documents

SRP6177

Sigma-Aldrich

CETP human

recombinant, expressed in insect cells, ≥90% (SDS-PAGE)

Synonym(s):

BPIFF, Cholesteryl ester transfer protein, HDLCQ10, Lipid transfer protein I

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in insect cells

Assay

≥90% (SDS-PAGE)

form

lyophilized powder

mol wt

55 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<1 EU/μg (LAL test)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CETP(1071)

General description

Cholesteryl ester transfer protein (CETP), also called plasma lipid transfer protein, is a plasma protein that facilitates the transport of cholesteryl esters and triglycerides between the lipoproteins. It collects triglycerides from very-low-density (VLDL) or low-density lipoproteins (LDL) and exchanges them for cholesteryl esters from high-density lipoproteins (HDL), and vice versa. Inhibition of CETP has been proposed as a strategy to raise HDL cholesterol levels, a potential an approach to retarding atherogenesis and a strategy to reduce cardiovascular risk.

Physical form

Lyophilized from a Buffer containing 10 mM Tris and 150 mM NaCl.

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

Reconstitute in sterile water at 2 mg/mL.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service