Skip to Content
Merck
All Photos(1)

Key Documents

SRP6080

Sigma-Aldrich

PPIL4 human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

Cyclophilin-like protein PPIL4, HDCME13P, Peptidyl prolyl cis-trans isomerase-like 4, Rotamase PPIL4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥85% (SDS-PAGE)

form

liquid

mol wt

59.4 kDa

packaging

pkg of 50 μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PPIL4(85313)

General description

PPIL4 is an evolutionarily conserved member of the cyclophilin-type PPIase family of proteins. Ubiquitously expressed with predominant expression in kidney, PPIL4 localizes to the nucleus and contains one PPIase cyclophilin-type domain, a lysine-rich domain, a pair of bipartite nuclear targeting sequences and one RRM (RNA recognition motif) domain. The presence of the RRM domain along with nuclear targeting sequences suggests that PPIL4 may be involved in transcriptional regulation.

Physical form

0.5 mg/mL solution in 20 mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2 mM DTT and 0.1 M NaCl.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MGSSHHHHHH SSGLVPRGSH MAVLLETTLG DVVIDLYTEE RPRACLNFLK LCKIKYYNYC LIHNVQRDFI IQTGDPTGTG RGGESIFGQL YGDQASFFEA EKVPRIKHKK KGTVSMVNNG SDQHGSQFLI TTGENLDYLD GVHTVFGEVT EGMDIIKKIN ETFVDKDFVP YQDIRINHTV ILDDPFDDPP DLLIPDRSPE PTREQLDSGR IGADEEIDDF KGRSAEEVEE IKAEKEAKTQ AILLEMVGDL PDADIKPPEN VLFVCKLNPV TTDEDLEIIF SRFGPIRSCE VIRDWKTGES LCYAFIEFEK EEDCEKAFFK MDNVLIDDRR IHVDFSQSVA KVKWKGKGGK YTKSDFKEYE KEQDKPPNLV LKDKVKPKQD TKYDLILDEQ AEDSKSSHSH TSKKHKKKTH HCSEEKEDED YMPIKNTNQD IYREMGFGHY EEEESCWEKQ KSEKRDRTQN RSRSRSRERD GHYSNSHKSK YQTDLYERER SKKRDRSRSP KKSKDKEKSK YR

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service