Skip to Content
Merck
All Photos(1)

Documents

SRP4972

Sigma-Aldrich

GHBP human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

GH receptor, GHBP, GHR, Somatotropin receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized

mol wt

~28.1 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GHR(2690)

General description

GHBP is a transmembrane receptor for growth hormone. Binding of growth hormone to the receptor leads to receptor dimerization and the activation of an intra- and intercellular signal transduction pathway leading to growth. A common alternate allele of this gene, called GHRd3, lacks exon three and has been well-characterized. Mutations in this gene have been associated with Laron syndrome, also known as the growth hormone insensitivity syndrome (GHIS), a disorder characterized by short stature. Human Recombinant GHBP expressed from E. coli is a single, non-glycosylated, polypeptide chain containing 237 amino acids and having a molecular mass of 28.1 kDa. GHBP is purified by proprietary chromatographic techniques.
The gene encoding growth hormone receptor (GHR or GHBP) is localized on human chromosome 5p13.1-p12, with nine exons. It possesses a cytoplasmic and an extracellular domain for protein binding.

Physical form

GHBP was lyophilized from a concentrated (1 mg/ml) solution with 0.0045 mM NaHCO3.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in sterile dH2O not less than 100 μg/ml. This solution can then be diluted into other aqueous buffers.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New molecular mechanisms of GH resistance
European Journal of Endocrinology, 151(1) (2004)
Growth hormone receptor (GHR) gene polymorphism and Prader--Willi syndrome.
Butler M G, et al.
American Journal of Medical Genetics, 161(7), 1647-1653 (2013)
De-Gang Song et al.
Oncotarget, 6(25), 21533-21546 (2015-06-24)
Chimeric antigen receptors (CARs) can redirect T cells against antigen-expressing tumors in an HLA-independent manner. To date, various CARs have been constructed using mouse single chain antibody variable fragments (scFvs) of high affinity that are immunogenic in humans and have
Eric L Carter et al.
The Journal of biological chemistry, 291(5), 2196-2222 (2015-12-17)
Rev-erbα and Rev-erbβ are heme-binding nuclear receptors (NR) that repress the transcription of genes involved in regulating metabolism, inflammation, and the circadian clock. Previous gene expression and co-immunoprecipitation studies led to a model in which heme binding to Rev-erbα recruits

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service