Skip to Content
Merck
All Photos(1)

Key Documents

SRP3009

Sigma-Aldrich

BAFF Receptor human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

BAFFR, TNFRSF13C

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
₪1,874.00

₪1,874.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
₪1,874.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

₪1,874.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

2.0-4.0 μg/mL ED50

mol wt

7.7 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

red to reddish

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

BAFF Receptor (BAFFR), a member of the TNFR superfamily, is highly expressed in spleen, lymph node, and resting B cells and to some extent in activated B cells, resting CD4+ cells and peripheral blood leukocytes. BAFFR is a type III transmembrane protein that binds with high specificity to BAFF (TNFSF13B). BAFFR/BAFF signaling plays a critical role in B cell survival and maturation. Recombinant human BAFFR is a 76 amino acid polypeptide (7.7 kDa) corresponding to the extracellular portion of the full BAFFR protein.

Biochem/physiol Actions

BAFF Receptor (BAFFR), a member of the TNFR superfamily, is highly expressed in spleen, lymph node, and resting B cells and to some extent in activated B cells, resting CD4+ cells and peripheral blood leukocytes. Recombinant human BAFFR is a 76 amino acid polypeptide (7.7 kDa) corresponding to the extracellular portion of the full BAFFR protein.

Sequence

MRRGPRSLRG RDAPAPTPCV PAECFDLLVR HCVACGLLRT PRPKPAGASS PAPRTALQPQ ESVGAGAGEA ALPLPG

Physical form

Lyophilized from 0.5x PBS, pH 7.2.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Waldschmidt et al.
Science (New York, N.Y.), 293(5537), 2012-2013 (2001-09-15)
What determines whether transitional B cells newly emerged from the bone marrow will differentiate further to become mature, long-lived, circulating B lymphocytes? In a Perspective, Waldschmidt and Noelle discuss new findings showing that the TNF family ligand BAFF and its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service