Skip to Content
Merck
All Photos(1)

Documents

SML1324

Sigma-Aldrich

XEN445

≥98% (HPLC)

Synonym(s):

(S)-2-(3-(Pyridin-2-ylmethoxy)pyrrolidin-1-yl)-5-(trifluoromethyl)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17F3N2O3
CAS Number:
Molecular Weight:
366.33
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C18H17F3N2O3/c19-18(20,21)12-4-5-16(15(9-12)17(24)25)23-8-6-14(10-23)26-11-13-3-1-2-7-22-13/h1-5,7,9,14H,6,8,10-11H2,(H,24,25)/t14-/m0/s1

InChI key

NBGRERFNOKZQLO-AWEZNQCLSA-N

Biochem/physiol Actions

XEN445 is a potent and selective inhibitor of endothelial lipase with an IC50 of 237 nM and high selectivity over hepatic and lipoprotein lipases. In experiments with wild-type mice, XEN445 raised plasma HDL-cholesterol levels by 16% after 3 days and by 30% after 9 days dosing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service