Skip to Content
Merck
All Photos(1)

Key Documents

SML1305

Sigma-Aldrich

T56-LIMKi

≥98% (HPLC)

Synonym(s):

3-Methyl-N-[3-[[[3-(trifluoromethyl)phenyl]amino]carbonyl]phenyl]-5-isoxazolecarboxamide, T5601640

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14F3N3O3
CAS Number:
Molecular Weight:
389.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC(C(F)(F)F)=CC=C1)C2=CC(NC(C3=CC(C)=NO3)=O)=CC=C2

InChI

1S/C19H14F3N3O3/c1-11-8-16(28-25-11)18(27)24-14-6-2-4-12(9-14)17(26)23-15-7-3-5-13(10-15)19(20,21)22/h2-10H,1H3,(H,23,26)(H,24,27)

InChI key

XVOKFRPKSAWELK-UHFFFAOYSA-N

Biochem/physiol Actions

T56-LIMKi is a LIMK2 inhibitor that reduces phosphorylated cofilin, and induces disassembly of actin stress fibers in NF1-/- MEFs cells.
T56-LIMKi is a LIMK2 inhibitor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service