Skip to Content
Merck
All Photos(1)

Key Documents

SMB00516

Sigma-Aldrich

Benzyl glucopyranoside

≥95% (LC/MS-ELSD)

Synonym(s):

benzyl D-glucopyranoside, benzyl-beta-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O6
CAS Number:
Molecular Weight:
270.28
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](O[C@@H]([C@H]([C@@H]1O)O)CO)OCC2=CC=CC=C2

InChI

1S/C13H18O6/c14-6-9-10(15)11(16)12(17)13(19-9)18-7-8-4-2-1-3-5-8/h1-5,9-17H,6-7H2/t9-,10-,11+,12-,13-/m1/s1

InChI key

GKHCBYYBLTXYEV-UJPOAAIJSA-N

General description

Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Mizuma et al.
Biochimica et biophysica acta, 1722(2), 218-223 (2005-02-18)
The intestinal absorption of benzyl beta-glucoside (BNZ beta glc) contained in the fruit of Prunus mume SIEB. et ZUCC. (Rosaceae), which is traditionally used as a medicinal food in Japan, was studied in rat intestines. BNZ beta glc was absorbed
Nobuhiko Fushimi et al.
Bioorganic & medicinal chemistry, 20(22), 6598-6612 (2012-10-16)
Sodium glucose co-transporter 1 (SGLT1) plays a dominant role in the absorption of glucose in the gut and is considered a promising target in the development of treatments for postprandial hyperglycemia. A series of 4-benzyl-1H-pyrazol-3-yl β-d-glucopyranoside derivatives have been synthesized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service