Skip to Content
Merck
All Photos(1)

Documents

SMB00350

Sigma-Aldrich

Dehydroandrographolide

≥98% (HPLC)

Synonym(s):

(1R-(1a,2a,4aa,5β(E),8aβ))-3-[2-(Decahydro-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylene-1-naphthalenyl)ethylidene)-2(3H)-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O4
CAS Number:
Molecular Weight:
332.43
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

plant (Andrographis paniculata)

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC[C@@]([C@H](O)CC1)(C)C([C@]1(C)[C@H]2C/C=C3C=COC\3=O)CCC2=C

InChI

1S/C20H28O4/c1-13-4-7-16-19(2,10-8-17(22)20(16,3)12-21)15(13)6-5-14-9-11-24-18(14)23/h5,9,11,15-17,21-22H,1,4,6-8,10,12H2,2-3H3/b14-5+/t15-,16?,17+,19-,20-/m0/s1

InChI key

YIIRVUDGRKEWBV-GJVBFGQCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dehydroandrographolide is a natural diterpenoid found in Andrographis paniculata.

Biochem/physiol Actions

Dehydroandrographolide possesses activity against hepatitis B virus DNA replication.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dehydroandrographolide enhances innate immunity of intestinal tract through up-regulation the expression of hBD-2
Xiong W B, et al.
DARU Journal of Pharmaceutical Sciences, 23(1), 37-37 (2015)
Naturally derived anti-hepatitis B virus agents and their mechanism of action
Wu Y H.
World Journal of Gastroenterology, 22(1), 188-188 (2016)
Synthesis, structure-activity relationships and biological evaluation of dehydroandrographolide and andrographolide derivatives as novel anti-hepatitis B virus agents.
Chen H, et al.
Bioorganic & Medicinal Chemistry Letters, 24(10), 2353-2359 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service