Skip to Content
Merck
All Photos(1)

Documents

SMB00325

Sigma-Aldrich

Triptonide

≥98% (HPLC)

Synonym(s):

Triptonide, 14-deoxy-14-oxo-triptolide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22O6
CAS Number:
Molecular Weight:
358.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O=C(OC1)C2=C1[C@]3([H])C[C@H]4[C@@]5(O4)C([C@]6(C(C)C)[C@@H](O6)[C@H](O7)[C@@]57[C@@]3(C)CC2)=O

InChI

1S/C20H22O6/c1-8(2)18-13(25-18)14-20(26-14)17(3)5-4-9-10(7-23-15(9)21)11(17)6-12-19(20,24-12)16(18)22/h8,11-14H,4-7H2,1-3H3/t11-,12-,13-,14-,17-,18-,19+,20+/m0/s1

InChI key

SWOVVKGLGOOUKI-ZHGGVEMFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triptonide is a toxic natural compound found in Tripterygium wilfordii.

Biochem/physiol Actions

Triptonide has potent antileukemic and antitumor activities.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service