SAB5702937
Anti-NOX2/gp91phox Antibody, clone 2Y8W3, Rabbit Monoclonal
Synonym(s):
AMCBX2,CGD,GP91-1,GP91-PHOX,GP91PHOX,IMD34,NOX2,p91-PHOX,CYBB
About This Item
Recommended Products
biological source
rabbit
Quality Level
material
colorless
clone
2Y8W3, monoclonal
form
liquid
mol wt
65 kDa
species reactivity
mouse, rat, human
concentration
0.62 mg/mL
technique(s)
western blot: 1:500 - 1:2000
color
colorless
isotype
IgG
immunogen sequence
FHKMVAWMIALHSAIHTIAHLFNVEWCVNARVNNSDPYSVALSELGDRQNESYLNFARKRIKNPEGGLYLAVTLLAGITGVVITLCLILIITSSTKTIRRS
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CYBB(1536)
General description
Immunogen
Application
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service