Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702479

Sigma-Aldrich

Anti-GABARAP Antibody, clone 5T6W5, Rabbit Monoclonal

Synonym(s):

ATG8A, GABARAP-a, MM46

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
₪1,725.00

₪1,725.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
₪1,725.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

₪1,725.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

5T6W5, monoclonal

form

liquid

mol wt

16 kDa

species reactivity

rat, rat, human

concentration

0.227mg/ml

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MKFVYKEEHPFEKRRSEGEKIRKKYPDRVPVIVEKAPKARIGDLDKKKYLVPSDLTVGQFYFLIRKRIHLRAEDALFFFVNNVIPPTSATMGQLYQEHHE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GABARAP(11337)

General description

Gamma-aminobutyric acid A receptors [GABA(A) receptors] are ligand-gated chloride channels that mediate inhibitory neurotransmission. This gene encodes GABA(A) receptor-associated protein, which is highly positively charged in its N-terminus and shares sequence similarity with light chain-3 of microtubule-associated proteins 1A and 1B. This protein clusters neurotransmitter receptors by mediating interaction with the cytoskeleton. [provided by RefSeq, Jul 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GABARAP (O95166).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service