Skip to Content
Merck
All Photos(3)

Key Documents

SAB5200049

Sigma-Aldrich

Monoclonal Anti-GABRB3 antibody produced in mouse

clone S87-25, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-GABA(A) receptor subunit beta3, Anti-GABAAR Beta3, S87-25, Anti-Gabrb3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
₪2,414.00

₪2,414.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
₪2,414.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

₪2,414.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S87-25, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 55 kDa

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Gabrb3(14402)

General description

The γ-aminobutyric acid type A receptor β3 subunit (GABRB3) gene with nine exons spanning 250kb, is mapped to human chromosome 15q12, a hot region of genomic rearrangements. The gene codes for β3 subunit of GABAA receptor, which belongs to the GABA (γ-aminobutyric acid) receptor gene family.

Specificity

Detects ~55 kDa. No cross-reactivity against GABA-A-R-Beta 2 or –Beta1.

Immunogen

Fusion protein amino acids 370-433 of mouse GABA-A_r Beta3, Accession Number AAB60502.

Biochem/physiol Actions

γ-aminobutyric acid type A receptor β3 subunit (GABRB3) is a candidate gene for autism spectrum conditions (ASC). Alteration in the gene results in increased tactile sensitivity, or hypersensitivity. Overexpression of GABRB3 might be implicated in the pathogenesis of heroin dependence. Aberration or mutation of this gene leads to neurodevelopmental disorders, such as Angelman syndrome, Prader-Willi syndrome and schizophrenia. GABRB3 polymorphisms results in nonsyndromic cleft lip and/or palate (NSCL/P).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in PBS, pH 7.4, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Hoerbelt et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(8), 3525-3536 (2015-02-27)
Dopamine is a critical neuromodulator that activates GPCRs in mammals or ligand-gated ion channels in invertebrates. The present study demonstrates that dopamine (0.1-10 mm) exerts novel, opposing effects on different populations of mammalian (rat) GABAA receptors. Using whole-cell patch-clamp electrophysiology
Teresa Tavassoli et al.
Molecular autism, 3(1), 6-6 (2012-07-10)
Autism spectrum conditions have a strong genetic component. Atypical sensory sensitivities are one of the core but neglected features of autism spectrum conditions. GABRB3 is a well-characterised candidate gene for autism spectrum conditions. In mice, heterozygous Gabrb3 deletion is associated
Chia-Hsiang Chen et al.
PloS one, 9(7), e102227-e102227 (2014-07-16)
GABRB3 encoding the β3 subunit of GABAA receptor has been implicated in multiple neuropsychiatric disorders, including substance abuse. Previous studies reported that SNPs at the 5' regulatory region of GABRB3 could regulate GABRB3 gene expression and associated with childhood absence
J Wagstaff et al.
American journal of human genetics, 49(2), 330-337 (1991-08-11)
Deletions of the proximal long arm of chromosome 15 (bands 15q11q13) are found in the majority of patients with two distinct genetic disorders, Angelman syndrome (AS) and Prader-Willi syndrome (PWS). The deleted regions in the two syndromes, defined cytogenetically and
K Glatt et al.
Genomics, 41(1), 63-69 (1997-04-01)
The genes encoding the gamma-aminobutyric acid (GABA) type-A receptor subunits beta 3 (GABRB3), alpha 5 (GABRA5), and gamma 3 (GABRG3) map to chromosome 15q11-q13. The three genes are contained within roughly 800 kb of the distal part of the imprinted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service