Skip to Content
Merck
All Photos(2)

Key Documents

SAB1409973

Sigma-Aldrich

Anti-MAP7 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

E-MAP-115, EMAP115

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
WB
clone:
polyclonal
species reactivity:
human
citations:
1
technique(s):
immunofluorescence: suitable
western blot: 1 μg/mL

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 84.1 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAP7(9053)

General description

The product of this gene is a microtubule-associated protein that is predominantly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis. (provided by RefSeq)

Immunogen

MAP7 (NP_003971.1, 1 a.a. ~ 749 a.a) full-length human protein.

Sequence
MAELGAGGDGHRGGDGAVRSETAPDSYKVQDKKNASSRPASAISGQNNNHSGNKPDPPPVLRVDDRQRLARERREEREKQLAAREIVWLEREERARQHYEKHLEERKKRLEEQRQKEERRRAAVEEKRRQRLEEDKERHEAVVRRTMERSQKPKQKHNRWSWGGSLHGSPSIHSADPDRRSVSTMNLSKYVDPVISKRLSSSSATLLNSPDRARRLQLSPWESSVVNRLLTPTHSFLARSKSTAALSGEAASCSPIIMPYKAAHSRNSMDRPKLFVTPPEGSSRRRIIHGTASYKKERERENVLFLTSGTRRAVSPSNPKARQPARSRLWLPSKSLPHLPGTPRPTSSLPPGSVKAAPAQVRPPSPGNIRPVKREVKVEPEKKDPEKEPQKVANEPSLKGRAPLVKVEEATVEERTPAEPEVGPAAPAMAPAPASAPAPASAPAPAPVPTPAMVSAPSSTVNASASVKTSAGTTDPEEATRLLAEKRRLAREQREKEERERREQEELERQKREELAQRVAEERTTRREEESRRLEAEQAREKEEQLQRQAEERALREREEAERAQRQKEEEARVREEAERVRQEREKHFQREEQERLERKKRLEEIMKRTRRTEATDKKTSDQRNGDIAKGALTGGTEVSALPCTTNAPGNGKPVGSPHVVTSHQSKVTVESTPDLEKQPNENGVSVQNENFEEIINLPIGSKPSRLDVTNSESPEIPLNPILAFDDEGTLGPLPQVDGVQTQQTAEVI

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brigette Y Monroy et al.
Developmental cell, 53(1), 60-72 (2020-02-29)
Many eukaryotic cells distribute their intracellular components asymmetrically through regulated active transport driven by molecular motors along microtubule tracks. While intrinsic and extrinsic regulation of motor activity exists, what governs the overall distribution of activated motor-cargo complexes within cells remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service