Skip to Content
Merck
All Photos(2)

Documents

SAB1400216

Sigma-Aldrich

Anti-SEPT5 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-CDCREL, Anti-CDCREL-1, Anti-CDCREL1, Anti-H5, Anti-PNUTL1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEPT5(5413)

General description

This gene is a member of the septin gene family of nucleotide binding proteins, originally described in yeast as cell division cycle regulatory proteins. Septins are highly conserved in yeast, Drosophila, and mouse and appear to regulate cytoskeletal organization. Disruption of septin function disturbs cytokinesis and results in large multinucleate or polyploid cells. This gene is mapped to 22q11, the region frequently deleted in DiGeorge and velocardiofacial syndromes. A translocation involving the MLL gene and this gene has also been reported in patients with acute myeloid leukemia. Two transcripts of this gene, a major one of 2.2 kb and a minor one of 3.5 kb, have been observed. The 2.2 kb form results from the utilization of a non-consensus polyA signal (AACAAT). In the absence of polyadenylation from this imperfect site, the consensus polyA signal of the downstream neighboring gene (GP1BB; platelet glycoprotein Ib) is used, resulting in the 3.5 kb transcript. An alternatively spliced transcript variant with a different 5′ end has also been identified, but its full-length nature has not been completely determined. (provided by RefSeq)

Immunogen

SEPT5 (NP_002679.2, 1 a.a. ~ 369 a.a) full-length human protein.

Sequence
MSTGLRYKSKLATPEDKQDIDKQYVGFATLPNQVHRKSVKKGFDFTLMVAGESGLGKSTLVHSLFLTDLYKDRKLLSAEERISQTVEILKHTVDIEEKGVKLKLTIVDTPGFGDAVNNTECWKPITDYVDQQFEQYFRDESGLNRKNIQDNRVHCCLYFISPFGHGLRPVDVGFMKALHEKVNIVPLIAKADCLVPSEIRKLKERIREEIDKFGIHVYQFPECDSDEDEDFKQQDRELKESAPFAVIGSNTVVEAKGQRVRGRLYPWGIVEVENQAHCDFVKLRNMLIRTHMHDLKDVTCDVHYENYRAHCIQQMTSKLTQDSRMESPIPILPLPTPDAETEKLIRMKDEELRRMQEMLQRMKQQMQDQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service