All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H14N2O6
CAS Number:
Molecular Weight:
234.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
−20°C
SMILES string
NC(CO)C(=O)NC(CCC(O)=O)C(O)=O
InChI
1S/C8H14N2O6/c9-4(3-11)7(14)10-5(8(15)16)1-2-6(12)13/h4-5,11H,1-3,9H2,(H,10,14)(H,12,13)(H,15,16)
InChI key
LAFKUZYWNCHOHT-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Laura Cipolla et al.
Future medicinal chemistry, 2(4), 587-599 (2011-03-24)
Several glycoprocessing enzymes and glycoreceptors have been recognized as important targets for therapeutic intervention. This concept has inspired the development of important classes of therapeutics, such as anti-influenza drugs inhibiting influenza virus neuraminidase, anti-inflammatory drugs targeting lectin-sialyl-Lewis X interaction and
T Tsukida et al.
Journal of medicinal chemistry, 40(22), 3534-3541 (1997-11-14)
We have rationally designed a sLe(x) mimetic based on molecular modeling, synthesized type II and type II' beta-turn dipeptides (3a,b), and evaluated their biological profiles both in vitro and in vivo. Against E-selectin-sLe(x) binding, the type II beta-turn dipeptide L-Ser-D-Glu
T Tsukida et al.
Journal of medicinal chemistry, 41(22), 4279-4287 (1998-10-23)
We have previously found that heterochiral fucodipeptides, L-Ser-D-Glu (3a) and D-Ser-L-Glu (3b), exhibited up to 20-100 times more potency than a sialyl Lewis X (sLeX, 1) and a 3'-sulfated Lewis X analogue (2) toward E-selectin binding and have also proposed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service