Skip to Content
Merck
All Photos(1)

Documents

S177

Sigma-Aldrich

5-HT7 Serotonin Receptor human

recombinant, expressed in CHO cells, membrane suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in CHO cells

Quality Level

form

membrane suspension

packaging

vial of 50 units

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HTR7(3363)

Physical form

Membrane suspension in 50 mM Tris-HCl at pH 7.4, 10% glycerol and 1% BSA. Actual concentrations and specific binding capacity are provided with each lot.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Bard et al.
The Journal of biological chemistry, 268(31), 23422-23426 (1993-11-05)
An intron-containing gene encoding a novel human serotonin (5-HT) receptor was isolated from human genomic and cDNA libraries with probes directed to transmembrane regions of the adenylate cyclase stimulatory Drosophila serotonin receptor gene, 5-HTdrol. Membranes harvested from transiently transfected Cos-7
M Ruat et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(18), 8547-8551 (1993-09-15)
By using a strategy based on nucleotide sequence homology, we have cloned a cDNA encoding a functional serotonin (5-HT) receptor. The deduced amino acid sequence of the 5-HT7 receptor displays limited homology with that of other 5-HT receptors. In addition
Y Shen et al.
The Journal of biological chemistry, 268(24), 18200-18204 (1993-08-25)
We have utilized the polymerase chain reaction technique to selectively amplify a G protein-coupled receptor cDNA from rat kidney proximal convoluted tubule mRNA, which exhibits high homology with previously cloned serotonin receptors. Sequencing of a full-length clone isolated from a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service