Skip to Content
Merck
All Photos(3)

Documents

RDP200

Sigma-Aldrich

GenElute FFPE RNA/DNA Purification Plus Kit

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600
NACRES:
NA.52

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

shipped in

wet ice

storage temp.

2-8°C

General description

Sigma′s GenElute FFPE RNA/DNA Purification Plus Kit provides a rapid method for the sequential isolation and purification of total RNA (including microRNA) and genomic DNA from formalin-fixed paraffin-embedded (FFPE) tissue samples. Using formalin to fix tissues leads to crosslinking of the nucleic acids and proteins, and the process of embedding the tissue samples can also lead to fragmentation of the nucleic acids over time. Sigma′s GenElute FFPE RNA/DNA Purification Plus Kit provides conditions that allow for the partial reversing of the formalin modifications, resulting in a high quality and yield of nucleic acids. The kit is able to purify all sizes of RNA, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA), depending on the age of the FFPE tissue as the degree of fragmentation of the RNA will increase over time. The RNA is purified from other cellular components without the use of phenol or chloroform. The purified RNA is of the highest integrity, and can be used in a number of downstream applications including real time PCR, reverse transcription PCR, Northern blotting, RNase protection and primer extension, and expression array assays. The purified genomic DNA is also of the highest quality, and can be used in PCR reactions, sequencing, Southern blotting and SNP analysis.

  • Fast and easy processing using rapid spin-column format
  • High yields and Quality of Nucleic Acids
  • Separate fractionation of RNA and DNA
  • Isolate total RNA, from large rRNA down to microRNA (miRNA)
  • No phenol or chloroform extractions

Application

RNA
  • Bioanalyzer
  • Quantitative, real-time RT-PCR for both large mRNA and small RNA including miRNA
  • RT-PCR for both large mRNA and small RNA including miRNA
  • Northern blotting
  • RNase protection
  • Primer extension
  • Expression array assays
  • Next Generation Sequencing
  • microRNA Cloning
  • miRNA array
  • RNA-Seq
  • Nanostring
  • Fluidigm
  • Droplet/Digital PCR


Genomic DNA
  • PCR and Quantitative PCR
  • Sequencing
  • DNA Methylation Studies
  • Southern blotting
  • Microarrays
  • SNP analysis

Features and Benefits

  • Complete column purification - Separate fractionation of RNA and DNA
  • Reduce variability - RNA and DNA are isolated from a single sample with no splitting of the lysate, thus reducing inconsistent results and variability
  • Isolate a diversity of RNA species - All sizes of RNA are isolated, from large mRNA down to microRNA
  • Isolate high quality RNA and DNA - The purified RNA and DNA are of the highest quality and can be used in a number of downstream applications

Unit Definition

Preps

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service