Skip to Content
Merck
All Photos(1)

Key Documents

R8257

Sigma-Aldrich

Mlu I from Micrococcus luteus (lysodeikticus)

buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Specificity

Recognition sequence: 5′-A/CGCGT-3′
Ligation and recutting results: After 2-10-fold Mlu I overdigestion of 1 μg λ DNA substrate, results in 100% cutting, >95% of fragments can be ligated, and >95% recut.
Heat inactivation: Partially inactivated at 65 °C for 10 minutes.

Other Notes

Supplied with 10x Restriction Endonuclease Buffer SH (B3657).

Caution

Comment: Mlu I will only partially cleave DNA isolated from E. coli strains that have the dam methylase (dam+ strains).

Physical form

Solution in 20 mM Tris-HCl, pH 8.0, 0.1 mM EDTA, 250 mM KCl, 10 mM 2-mercaptoethanol, 50% glycerol (v/v), 0.2% Triton X-100 (v/v) at 4°C

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Sugisaki et al.
Gene, 16(1-3), 73-78 (1981-12-01)
Two new restriction endonucleases have been isolated from Flavobacterium okeanokoites IFO12536 and Micrococcus luteus IFO12992 and named FokI and MluI, respectively. Based on analysis of the sequences around the restriction sites, the recognition sequences and cleavage sites of these endonucleases
C Kessler et al.
Gene, 92(1-2), 1-248 (1990-08-16)
The properties and sources of all known class-I, class-II and class-III restriction endonucleases (ENases) and DNA modification methyltransferases (MTases) are listed and newly subclassified according to their sequence specificity. In addition, the enzymes are distinguished in a novel manner according
Nico Mitro et al.
Methods in molecular biology (Clifton, N.J.), 952, 137-144 (2012-10-27)
The role of certain amino acids in the interactions of ligands with their cognate nuclear receptors is usually achieved by the resolution of the crystal structure of the receptor complexed with the ligand. As a complementary functional approach, site-directed mutagenesis
Rachel M Smith et al.
Nucleic acids research, 41(1), 391-404 (2012-11-14)
Type IIB restriction-modification systems, such as BcgI, feature a single protein with both endonuclease and methyltransferase activities. Type IIB nucleases require two recognition sites and cut both strands on both sides of their unmodified sites. BcgI cuts all eight target
Lilia Gabsalilow et al.
Nucleic acids research, 41(7), e83-e83 (2013-02-15)
Targeted genome engineering requires nucleases that introduce a highly specific double-strand break in the genome that is either processed by homology-directed repair in the presence of a homologous repair template or by non-homologous end-joining (NHEJ) that usually results in insertions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service