Skip to Content
Merck
All Photos(1)

Key Documents

R2529

Sigma-Aldrich

Anti-p190 RhoGAP (DG-20) antibody produced in rabbit

~3 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GRLF1, Anti-Glucorticoid Receptor Binding Factor 1, Anti-p190A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 190 kDa

species reactivity

rat

concentration

~3 mg/mL

technique(s)

immunoprecipitation (IP): 40-60 μg using p190 RhoGAP protein from a MDCK cell lysate
western blot: 2-4 μg/mL using mouse brain extract (S1 fraction), a whole cell extract of the Madin-Darby canine kidney MDCK cell line or the HeLa human epitheloid carcinoma cell line.

shipped in

dry ice

storage temp.

−20°C

Gene Information

Immunogen

synthetic peptide corresponding to amino acids 1181-1200 located near the C-terminus of rat p190 RhoGAP, conjugated to KLH.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service